Skip to Content
Merck
All Photos(1)

Key Documents

8.08308

Sigma-Aldrich

Titanium(III) chloride solution

about 30%, in about 10% hydrochloric acid, for synthesis

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€76.00
500 ML
€286.00
50 KG
€15,280.00

€76.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€76.00
500 ML
€286.00
50 KG
€15,280.00

About This Item

UNSPSC Code:
12352302
NACRES:
NA.22

€76.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Product Name

Titanium(III) chloride solution about 30%, (in about 10% hydrochloric acid) for synthesis

Quality Level

form

solution

density

1.334 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Assay (cerimetric TiCl₃): 27 - 33 %

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How is 30% TiCl3 solution synthesized and how does this synthesis compare with 20% solution in HCl?

    1 answer
    1. Unfortunately, the manufacturing process of this product and 20% solution in HCl are both considered proprietary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service