Skip to Content
Merck
All Photos(1)

Documents

1.10789

Sigma-Aldrich

Titanium(III) chloride solution

15%, (in about 10% hydrochloric acid) for amino acid analysis Reag. Ph Eur

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.21

product name

Titanium(III) chloride solution about 15%, (in about 10% hydrochloric acid) for amino acid analysis Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

form

liquid

pH

<1 (20 °C in H2O, undiluted)

density

1.21 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Assay
- cerimetric TiCl₃:
14.5 - 15.5 %
- argentometric, HCl: 9.0 - 11.0 %
Density (d 20/20): about 1.19
Sulphate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.005 %
Fe (Iron): ≤ 0.005 %

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service