Skip to Content
Merck
All Photos(1)

Key Documents

8.07574

Sigma-Aldrich

2-Pyrrolecarbaldehyde

for synthesis

Synonym(s):

2-Pyrrolecarbaldehyde, Pyrrole-2-aldehyde, Pyrrole-2-carboxaldehyde, 2-Formylpyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO
CAS Number:
Molecular Weight:
95.10
MDL number:
UNSPSC Code:
12352114
EC Index Number:
213-705-5
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

bp

217-219 °C

mp

43-46 °C

transition temp

flash point >100 °C

InChI

1S/C5H5NO/c7-4-5-2-1-3-6-5/h1-4,6H

InChI key

ZSKGQVFRTSEPJT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 43 °C
Melting range (upper value): ≤ 46 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service