Skip to Content
Merck
All Photos(2)

Key Documents

130052

Sigma-Aldrich

N-Methyl-2-pyrrolecarboxaldehyde

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€74.40
25 G
€194.00

€74.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€74.40
25 G
€194.00

About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
Beilstein:
107811
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€74.40


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

bp

87-90 °C/22 mmHg (lit.)

density

1.016 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)c1cccn1C

InChI

1S/C6H7NO/c1-7-4-2-3-6(7)5-8/h2-5H,1H3

InChI key

OUKQTRFCDKSEPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Methyl-2-pyrrolecarboxaldehyde was used to prepare 3-(1-methyl-1H-pyrrol-2-yl)propanoic acid[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Luo et al.
Bioorganic & medicinal chemistry, 18(14), 5048-5055 (2010-07-20)
A series of novel benzimidazole derivatives were synthesized and evaluated for their anti-hepatitis B virus (HBV) activity and cytotoxicity in the HepG2.2.15 cell line. The preliminary SAR was discussed. Compound 12a, with IC50<0.41 microM and SI>81.2, was the most promising

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service