Skip to Content
Merck
All Photos(1)

Key Documents

8.03548

Sigma-Aldrich

Dipropylamine

for synthesis

Synonym(s):

Dipropylamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€28.50
500 ML
€34.50
1 L
€45.70
20 KG
€514.00

€28.50


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€28.50
500 ML
€34.50
1 L
€45.70
20 KG
€514.00

About This Item

Linear Formula:
(CH3CH2CH2)2NH
CAS Number:
Molecular Weight:
101.19
MDL number:
UNSPSC Code:
12352116
EC Index Number:
205-565-9
NACRES:
NA.22

€28.50


Estimated to ship on07 April 2025Details


Request a Bulk Order

vapor pressure

38 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

260 °C

potency

925 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-9.3 % (v/v)

bp

105 °C/1013 hPa

mp

-63 °C

transition temp

flash point 7 °C

solubility

35 g/L (experimental)

density

0.74 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCC)CCC

InChI

1S/C6H15N/c1-3-5-7-6-4-2/h7H,3-6H2,1-2H3

InChI key

WEHWNAOGRSTTBQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.738 - 0.740
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service