Skip to Content
Merck
All Photos(1)

Key Documents

8.07483

Sigma-Aldrich

Polyethylene glycol

200, for synthesis

Synonym(s):

Poly(ethylene glycol), Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 200, PEG

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€62.20
5 L
€213.00

€62.20


Estimated to ship on11 June 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€62.20
5 L
€213.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
NACRES:
NA.22

€62.20


Estimated to ship on11 June 2025Details


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

350 °C (DIN 51794)

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

5-7 (20 °C, 100 g/L in H2O)

bp

>150 °C

mp

-50 °C

transition temp

flash point 180 °C

solubility

soluble 70 g/L

density

1.124 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyethylene glycol 200 can be used as:
  • A structure-directing agent to synthesize iron vanadate (FeVO4) nanoparticles via co-precipitation method.[1]
  • An organic additive to prepare crystalline nanorods of calcium tungstate (CaWO4) using calcium chloride and sodium tungstate via solvothermal process.[2]
  • A surface modifier in the synthesis of calcium peroxide nanoparticles using CaCl2 as a precursor via hydrolysis-precipitation method.[3]
  • A green solvent in combination with H2O in the preparation of 4-sulfanylcoumarins by sulfanylation of 4-tosyloxycoumarins with thiourea and alkyl halides.[4]

Analysis Note

Density (d 20 °C/ 4 °C): 1.124 - 1.126
Hydroxyl value: 535 - 590
Average molecular mass: 190 - 210
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. IS YOUR PEG 200 USP PHARMACEUTICAL GRADE?

    1 answer
    1. This item is intended for research use only and is not suitable for pharmaceutical manufacturing. Please see the following links below to review products that are pharmaceutical grade options:
      https://www.sigmaaldrich.com/product/mm/817001
      https://www.sigmaaldrich.com/product/mm/817005
      https://www.sigmaaldrich.com/product/mm/817019
      https://www.sigmaaldrich.com/product/mm/817045

      Helpful?

  4. What is the concentration of this product?

    1 answer
    1. This product is a neat liquid considered to be a 100% concentration.

      Helpful?

  5. What is the concentration of this product?

    1 answer
    1. This product a neat liquid considered to be a 100% concentration.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service