Skip to Content
Merck
All Photos(1)

Key Documents

8.03211

Sigma-Aldrich

Decanal

for synthesis

Synonym(s):

Decanal, Capraldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€57.20

€57.20


Estimated to ship on10 March 2025Details



Select a Size

Change View
100 ML
€57.20

About This Item

Linear Formula:
CH3(CH2)8CHO
CAS Number:
Molecular Weight:
156.27
MDL number:
UNSPSC Code:
12352114
EC Index Number:
203-957-4

€57.20


Estimated to ship on10 March 2025Details


vapor pressure

0.1 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

autoignition temp.

200 °C

potency

3096 mg/kg LD50, oral (Rat)
4183 mg/kg LD50, skin (Rabbit)

bp

207-209 °C/1013 hPa

mp

7 °C

transition temp

flash point 87 °C

density

0.83 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=CCCCCCCCCC

InChI

1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h10H,2-9H2,1H3

InChI key

KSMVZQYAVGTKIV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.826 - 0.829
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service