Skip to Content
Merck
All Photos(2)

Key Documents

1.03853

Millipore

GranuCult® prime Salmonella Shigella (SS) Agar

ISO 6579

Synonym(s):

SS Agar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€190.00

€190.00


Estimated to ship on07 April 2025Details


Request more information

Select a Size

Change View
500 G
€190.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€190.00


Estimated to ship on07 April 2025Details


Request more information

Agency

ISO 6579

Quality Level

form

powder

technique(s)

microbiological culture: suitable

pH

7.0±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

selective and differential for Salmonella Shigella

General description

GranuCult® prime Salmonella Shigella (SS) Agar is a nutrient rich medium which is designed to isolate and differentiate Salmonella spp. from food, animal feed, water, and other test samples.
The presence of brilliant green, ox bile, high concentrations of thiosulfate and citrate largely inhibits the accompanying flora and swarming of Proteus spp, while allowing the growth of Salmonella spp. The synergistic reaction between thiosulfate and iron salt present in the media enables the growth of H2S positive black color colonies.
Lactose present in the media differentiates between the lactose positive and lactose negative microorganisms. Lactose fermenting microbes form an acid which is indicated by forming red colonies due to the presence of a pH indicator, neutral red. Peptones provide nitrogen, vitamins, amino acids, and minerals for growth and agar acts as a solidifying agent.

Application

GranuCult® prime Salmonella Shigella (SS) Agar is used to isolate and differentiate Salmonella spp. and Shigella spp from food, animal feed, water, and other test samples.

Analysis Note

  • Salmonella

  • spp. produce translucent colonies with or without black center and yellowish-brown colored medium or may appear as almost black colonies due to H2S production.
  • Shigella spp. produce translucent colonies without black center and yellowish-brown colored medium.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service