Skip to Content
Merck
All Photos(1)

Key Documents

8.01221

Sigma-Aldrich

Ammonium sulfite

(35% solution in water) for synthesis

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€20.60
2.5 L
€35.60
25 L
€140.00
200 L
€953.00

€20.60


Estimated to ship on10 June 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€20.60
2.5 L
€35.60
25 L
€140.00
200 L
€953.00

About This Item

UNSPSC Code:
12352301
NACRES:
NA.22

€20.60


Estimated to ship on10 June 2025Details


Request a Bulk Order

Quality Level

form

liquid

pH

7.5-7.6 (20 °C in H2O)

mp

4-8 °C

density

1.19 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (iodometric): 37.0 - 40.0 %
Density (d 20 °C/ 4 °C): 1.195 - 1.205

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service