Skip to Content
Merck
All Photos(1)

Key Documents

1.05442

Supelco

Ammonium sulfide solution

GR for analysis

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€231.00

€231.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€231.00

About This Item

UNSPSC Code:
12352301
NACRES:
SE.11

€231.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

vapor pressure

450 hPa ( 20 °C)

Quality Level

form

liquid

expl. lim.

4-46 % (v/v)

pH

9.5 (450 g/L in H2O)

transition temp

flash point 15 °C

density

0.997 g/cm3 at 20 °C

storage temp.

no temp limit

Application


  • Improved Anti-Vulcanization and Bonding Performance of a Silver Alloy Bonding Wire by a Cathodic Passivation Treatment with Palladium: This study highlights the application of ammonium sulfide solution in enhancing the anti-vulcanization and bonding properties of silver alloy bonding wires used in electronic devices. The cathodic passivation treatment using ammonium sulfide offers significant improvements in the durability and performance of materials, which is pivotal in material science and industrial manufacturing (Yang et al., 2022).

  • The roles and applications of chaotropes and kosmotropes in industrial fermentation processes: Ammonium sulfide, utilized as a chaotrope, plays a crucial role in stabilizing enzymes and proteins during fermentation processes. This application is particularly important in biotechnological and pharmaceutical industries, where enzyme stability is critical to the efficiency of bioprocesses (Timson, 2020).

  • Cobalt iontophoresis techniques for tracing afferent and efferent connections in the vertebrate CNS: While primarily focused on cobalt iontophoresis, this research outlines the potential utility of ammonium sulfide in similar neuroscientific applications. Ammonium sulfide can be used for tracing neural connections due to its effective ion transport properties, providing valuable insights into neural pathway studies (Fuller and Prior, 1975).

Analysis Note

Assay (as (NH₄)₂S from sulphide): ≥ 20 %
Chloride (Cl): ≤ 0.05 %
Ca (Calcium): ≤ 0.003 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.001 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Pb (Lead): ≤ 0.0005 %
Residue on ignition (800 °C as sulfate): ≤ 0.05 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F

Flash Point(C)

15 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service