Skip to Content
Merck
All Photos(1)

Key Documents

71353-M

Sigma-Aldrich

Rosetta-gami 2(DE3)pLacI Competent Cells - Novagen

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing

Select a Size

0.4 ML
€213.00
1 ML
€395.00

€213.00


Estimated to ship on03 March 2025Details



Select a Size

Change View
0.4 ML
€213.00
1 ML
€395.00

About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

€213.00


Estimated to ship on03 March 2025Details


Product Name

Rosetta-gami 2(DE3)pLacI Competent Cells - Novagen, Rosetta-Gami 2 strains allows for enhanced disulfide bond formation and enhanced expression of eukaryotic proteins. Contains the pLacI plasmid producing extra Lac repressor.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: 2×106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: Δ(ara-leu)7697 ΔlacX74 ΔphoA PvuII phoR araD139 ahpC galE galK rpsL (DE3) F′[lac+ lacIq pro] gor522::Tn10 trxB pLacIRARE2 (CamR, StrR, TetR)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Expression host strains for pETBlue and pTriEx vectors are lDE3 lysogens that also carry the compatible pLacI plasmid, encoding the lac repressor. The pLacI plasmid confers chloramphenicol resistance and provides enough lac repressor to inhibit transcription from the T7lac promoter in the absence of inducer. The NovaBlue host strain is recommended for initial cloning with pETBlue and pTriEx vectors due to its superior transformation efficiency and high yields of high-quality plasmid DNA.

Note that these strains are not recommended for use with pET, pACYCDuet, pCDF, pRSF, pCOLADuet or pETcoco vectors.

.Rosetta-gami 2 host strains combine the advantages of Rosetta 2 and Origami 2 strains to alleviate codon bias and enhance disulfide bond formation in the cytoplasm when heterologoous proteins are expressed in E. coli. These trxB/gor mutants are compatible with kanamycin-resistant vectors, and carry the chloramphenicol-resistant pRARE2 plasmid, which supplies seven rare tRNAs. In Rosetta-gami 2(DE3)pLacI, the rare tRNA genes are present on the same plasmid that carries the lac repressor gene.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
Rosetta-Gami 2 strains allows for enhanced disulfide bond formation and enhanced expression of eukaryotic proteins. Contains the pLacI plasmid producing extra Lac repressor.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlRosetta-gami 2(DE3)pLacI Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•2 ng2 ngTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service