Yes, the Origami 2 host strains have deletion mutations in both the thioredoxin reductase (trxB) and glutathione reductase (gor) genes resulting in a natural expression. These are permanant deletions which do not require the use of antibiotics to maintain expression. Please see the link below to review additional information:
https://www.emdmillipore.com/product/Origami-2DE3pLysS-Competent-Cells-Novagen,EMD_BIO-71346#anchor_USP
71346-M
Origami 2(DE3)pLysS Competent Cells - Novagen
Escherichia coli, rod shaped
Synonym(s):
Novagen Competent Cells
About This Item
Recommended Products
Product Name
Origami 2(DE3)pLysS Competent Cells - Novagen, Origami 2 strains have mutations in glutathione reducatase(gor) and thioredoxin reductase (trxB), facilitating proper disulfide bond formation. T7 lysozyme expression suppresses basal T7 expression.
biological source
Escherichia coli
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze
growth mode
adherent or suspension
morphology
rod shaped
technique(s)
microbiological culture: suitable
cell transformation
transformation efficiency: 2×106 cfu/μg
shipped in
dry ice
storage temp.
−70°C
Related Categories
General description
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Components
•2 × 0.2 ml5 × 0.2 mlOrigami 2(DE3)pLysS Competent Cells
•2 × 2 ml4 × 2 mlSOC Medium
•2 ng2 ngTest Plasmid
Warning
Legal Information
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is the trxB gene naturally expressed in Origami 2 cells? If it is not naturally expressed, what antibiotics are required for expression?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service