Skip to Content
Merck
All Photos(1)

Key Documents

70188-M

Sigma-Aldrich

pSTBlue-1 Blunt Vector (linearized) - Novagen

Sign Into View Organizational & Contract Pricing

Select a Size

20 REACTIONS
€260.00

€260.00


Estimated to ship on24 March 2025Details



Select a Size

Change View
20 REACTIONS
€260.00

About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

€260.00


Estimated to ship on24 March 2025Details


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−70°C

General description

The pSTBlue-1 vector is a multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, dual opposed T7/SP6 promoters and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3′ overhangs are conveniently positioned near each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease III.

For enhanced convenience, please consider Introductory pSTBlue-1 Perfectly Blunt®Cloning Kit, or pT7Blue-1 Perfectly Blunt Cloning Kit.

Components

20 rxn40 rxn

•2 × 0.5 µg4 × 0.5 µgBlunt Vector

•10 µl10 µlPositive Control Insert

•100 µl2 × 100 µlEnd Conversion Mix

Warning

Toxicity: Standard Handling (A)

Other Notes

Novy, R.E., et al. 1996. inNovations6, 7. Clark, J.M. 1988. Nucleic Acids Res. 16, 9677. Brownstein, J.M., et al. 1996. BioTechniques20, 1004. Magnuson, V.L., et al. 1996. BioTechniques21, 700. Hu, G. 1993. DNA and Cell Biology12, 763.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PT7BLUE is a trademark of Merck KGaA, Darmstadt, Germany
Perfectly Blunt is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service