Skip to Content
Merck
All Photos(1)

Key Documents

5.43833

Supelco

Sodium acetate trihydrate

for HPLC LiChropur

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€130.00
250 G
€283.00

€130.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€130.00
250 G
€283.00

About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
MDL number:
UNSPSC Code:
41116105
EC Index Number:
204-823-8
NACRES:
NB.21

€130.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Quality Level

form

solid

autoignition temp.

1112 °F
607 °C

technique(s)

HPLC: suitable

pH

7.5-9.2 (20 °C, 50 g/L in H2O)

mp

58 °C

transition temp

flash point >250 °C

density

1.42 g/cm3 at 20 °C

bulk density

900 kg/m3

storage temp.

2-30°C

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • 3D Printing of Supercooled Liquids: Sodium acetate trihydrate is used in 3D printing of supercooled liquids. This research involves modeling and verifying the behavior of supercooled sodium acetate trihydrate in additive manufacturing processes. The study demonstrates its potential in creating complex structures and highlights the thermal properties essential for maintaining supercooling during the printing process. This application is significant for chemists working on advanced manufacturing techniques and material science (Brillinger, Additive Manufacturing, 2023).

  • Thermal Energy Storage: Sodium acetate trihydrate is widely used in thermal energy storage systems due to its favorable phase change properties. Studies have shown its effectiveness in storing and releasing thermal energy, which is crucial for applications in solar heating and building energy efficiency. The research also explores the impact of adding water to sodium acetate trihydrate to enhance its thermal stability and latent heat capacity (Beaupere et al., Applied Sciences, 2021).

  • Electrical Nucleation of Supercooled Phase Change Composites: Sodium acetate trihydrate is used in the preparation and performance analysis of controllably-triggered electrodes for the electrical nucleation of supercooled phase change composites. This study focuses on improving the efficiency and reliability of phase change materials for thermal management systems. The findings are crucial for developing advanced thermal storage solutions in various industrial applications (Chen et al., SSRN Electronic Journal, 2022).

  • Residual Solidification Duration Measurements: Experimental measurements of the residual solidification duration of supercooled sodium acetate trihydrate are conducted to understand its phase change dynamics. This research provides insights into the kinetics of solidification and helps optimize the material′s use in thermal energy storage applications. The study is vital for chemists working on phase change materials and energy storage systems (Beaupere et al., International Journal of Thermal Sciences, 2020).

  • Composite Phase Change Materials: Sodium acetate trihydrate is combined with pyrolytic carbon residue to create composite phase change materials. This research aims to improve the thermal cycling performance and reduce the undercooling and phase separation issues commonly associated with sodium acetate trihydrate. The study demonstrates the material′s stability and high latent heat value, making it suitable for long-term thermal energy storage applications (Yuan, Journal of Physics: Conference Series, 2022).

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service