Skip to Content
Merck
All Photos(1)

Key Documents

539141

Sigma-Aldrich

Proteasome Substrate II, Fluorogenic

Fluorogenic proteasome substrate.

Synonym(s):

Proteasome Substrate II, Fluorogenic, Z-LLE-AMC

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€233.00

€233.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
5 MG
€233.00

About This Item

Empirical Formula (Hill Notation):
C35H44N4O9
Molecular Weight:
664.75
UNSPSC Code:
12352200
NACRES:
NA.77

€233.00


Estimated to ship on22 April 2025Details


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

fluorescence

λex ~380 nm
λem ~460 nm

shipped in

ambient

storage temp.

−20°C

General description

Fluorogenic proteasome substrate (excitation maximum: 380 nm; emission maximum: 460 nm).
Fluorogenic proteasome substrate.

Specificity

Inhibits serine proteases, cysteine proteases. Does not inhibit metalloproteases.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Fluorogenic proteasome substrate
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Leu-Leu-Glu-AMC

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service