Skip to Content
Merck
All Photos(1)

Key Documents

208772

Sigma-Aldrich

Calpain-1 Substrate II, Fluorogenic

An internally quenched peptide substrate that is optimized for calpain-1 amino acid recognition motifs at the primed as well as the non-primed sides.

Synonym(s):

Calpain-1 Substrate II, Fluorogenic, H-E(EDANS)PLF~AERK(DABCYL)-OH

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€267.00

€267.00


Estimated to ship on05 May 2025Details



Select a Size

Change View
1 MG
€267.00

About This Item

Empirical Formula (Hill Notation):
C72H97N17O16S
Molecular Weight:
1488.71
UNSPSC Code:
12352200
NACRES:
NA.77

€267.00


Estimated to ship on05 May 2025Details


Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

red

solubility

DMSO: 5 mg/mL

fluorescence

λex ~505 nm
λem ~335 nm

shipped in

wet ice

storage temp.

−20°C

General description

An internally quenched peptide substrate that is optimized for calpain-1 amino acid recognition motifs at the primed as well as the non-primed sides. Shown to be kinetically superior, in terms of turnover rate, to Calpain Substrate III, Fluorogenic (Cat. No. 208771), Calpain-1 Substrate, Fluorogenic (Cat. No. 208748), and Calpain Substrate II, Fluorogenic (Cat. No. 208731) by ~8-, 18-, and 310-fold, respectively. Excitation: ~335 nm; emission: ~505 nm.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Calpain-1 Substrate
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Sequence

H₂N-Glu(EDANS)-Pro-Leu-Phe~Ala-Glu-Arg-Lys(DABCYL)-CO₂H

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cuerrier, D., et al. 2007. J. Biol. Chem.282, 9600.
Cuerrier, D., et al. 2005. J. Biol. Chem.280, 40632.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service