Skip to Content
Merck
All Photos(1)

Key Documents

5.33642

Sigma-Aldrich

AMP Deaminase Inhibitor, Cpd3

Synonym(s):

AMP Deaminase Inhibitor, Cpd3, (S)-6-(4-(((1-(isoquinolin-8-yl)ethyl)amino)methyl)phenyl)nicotinic acid , AMPD Inhibitor, Cpd3 , AMPD Inhibitor, AMP Deaminase Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€217.00

€217.00


Estimated to ship on21 May 2025Details



Select a Size

Change View
5 MG
€217.00

About This Item

Empirical Formula (Hill Notation):
C24H21N3O2
Molecular Weight:
383.44
UNSPSC Code:
12352200
NACRES:
NA.77

€217.00


Estimated to ship on21 May 2025Details


Assay

≥98% (HPLC)

Quality Level

form

solid

potency

38 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable isoquinoline derivative that acts as a specific AMP competitive inhibitor of AMP deaminase (IC50 = 38, 27, and 24 nM for human, rat, and mice, respectively). Potentiates low frequency electrical stimulation induced increase in AMP level, AMP:ATP ratio, and phosphorylation of AMPK (at Thr172), and acetyl-CoA carboxylase (at Ser218) in contracting rat epitrochlearis muscle. However, it does not affect basal AMPK activity and contraction-stimulated glucose uptake. Preincubation with this compound reduces basal IMP levels, but does affect the contraction-induced increases in IMP, adenosine, and inosine in any appreciable manner.

Biochem/physiol Actions

Cell permeable: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Admyre, T., et al. 2014. Chem. Biol.21, 1486.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What food contains this substance

    1 answer
    1. Unfortunately, a list of foods containing this substance can not provided.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service