Skip to Content
Merck
All Photos(2)

Key Documents

116860

Sigma-Aldrich

Adenosine Deaminase Inhibitor, DCF

The Adenosine Deaminase Inhibitor, DCF, also referenced under CAS 53910-25-1, controls the biological activity of Adenosine Deaminase.

Synonym(s):

Adenosine Deaminase Inhibitor, DCF, 2ʹ-Deoxycoformycin, (8R)-3-(2-deoxy-β-D-erythro- pentofuranosyl)-3,6,7,8-tetrahydroimidazo[4,5-d][1,3]diazepin-8-ol, Pentostatin, S. antibioticus

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€176.00

€176.00


Estimated to ship on03 February 2025Details



Select a Size

Change View
10 MG
€176.00

About This Item

Empirical Formula (Hill Notation):
C11H16N4O4
CAS Number:
Molecular Weight:
268.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

€176.00


Estimated to ship on03 February 2025Details


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL, clear, colorless
water: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

O[C@@H]1CNC=NC2=C1N=CN2[C@@H]3O[C@H](CO)[C@H](O)C3[H]

InChI

1S/C11H16N4O4/c16-3-8-6(17)1-9(19-8)15-5-14-10-7(18)2-12-4-13-11(10)15/h4-9,16-18H,1-3H2,(H,12,13)/t6-,7+,8+,9+/m0/s1

InChI key

FPVKHBSQESCIEP-JQCXWYLXSA-N

General description

A cell-permeable purine nucleoside compound that acts as a highly potent, tight binding transition state analog inhibitor of adenosine deaminase (ADA; Ki= 2.5 pM against partially purified human erythrocytes, and 0.48 to 9.1 nM against rat liver, rat intestine, rat hepatoma and human B cells). Reported to be an immunomodulator and exert selective toxicity towards lymphocytes.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Robak, T. 2007. Cancer Treat. Rev.33, 710.
Klohs, W.D. and Kraker, A.J., 1992. Pharmacol. Rev.44, 459.
Agarwal, R.P., et al. 1977. Biochem. Pharmacol.26, 359.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service