Skip to Content
Merck
All Photos(1)

Key Documents

5.31662

Sigma-Aldrich

JHDM Inhibitor VIII, SD70

Synonym(s):

JHDM Inhibitor VIII, SD70, N-(Furan-2-yl(8-hydroxyquinolin-7-yl)methyl)isobutyramide, N-(2-Furyl(8-hydroxy-7-quinolinyl)methyl)-2-methylpropanamide, Histone Lysine Demethylase Inhibitor XV, SD-70

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€205.00

€205.00


Estimated to ship on22 May 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C18H18N2O3
CAS Number:
Molecular Weight:
310.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on22 May 2025Details


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

N(C(c3[o]ccc3)c1c(c2ncccc2cc1)O)C(=O)C(C)C

InChI key

PBZYAPZLINAHET-UHFFFAOYSA-N

General description

A cell-permeable compound with a Fe(II)-chelating 8-hydroxyquinoline (8HQ) moiety is reported to inhibit KDM4C (IC50 = 30 µM against calf thymus histone H3K9me2 demethylation; [FeSO4] = 50 µM & [α-ketoglutarate] = 1 mM) and possibly other JMJD2/KDM4 members of Jumonji domain-containing histone demethylases (JHDM). Effectively suppresses androgen receptor (AR) target genes expression in human prostate adenocarcinoma LNCaP cultures upon 100 nM DHT stimulation as well as in CWR22Rv1 cells with constitutively active AR (10 µM) without affecting topoisomerase activity of E. coli Topo I or human Topo II. Shown to prevent AR/genotoxic stress-induced TMPRSS2 chromosomal translocation and gene fusions in LNCaP cells (1/100 & 1/13 of control TMPRSS2ETV1 and TMPRSS2ERG induction level, respectively; 10 µM SD70 dosed 2 h prior to 24 h 50 Gy irradiation & 100 nM DHT) and exhibit anti-proliferation activity against CWR22Rv1 both in cultures (97% decreased vs. 260% increased viable cell number on d6 by MTS assay with or without 10 µM SD70 ) in vitro and in mice (84% growth retardation on d10; 10 mg/4 mL/kg/d i.p.) in vivo without apparent animal toxicity. LNCaP genomic binding sites analyses utilizing Chem-seq with Biotin-derivatized SD70 as well as antibody-based ChIP reveal that 46% of genes upregulated by >1.5-fold upon DHT stimulation have both AR enhancer and SD70-binding sites within 500 kb of the coding gene promoter and 42% of all SD70-suppressed genes in DHT-treated LNCaP cells are DHT-induced genes. Likewise, increased KDM4C binding upon DHT treatment is observed on AR enhancers coocuppied by SD70.
A cell-permeable, bioavailable, non-toxic 8-hydroxyquinolinyl based compound that binds to and colocalizes with androgen receptor (AR) at both ligand-dependent and independent transcriptional regulatory elements. Acts as an inhibitor of dihydotestosterone-regulated gene expression. Shown to inhibit prostate cancer cell transcription by blocking the activity of histone lysine demethylase KDM4C (IC50 = 30 µM). Increases total H3K9me2 levels in 293 T cells in enhancer and promoter regions and causes a slight diminution in the levels of methylated H3K36. Does not affect the activity of DNA/RNA polymerase or topoisomerase I and II activities in any significant manner. Blocks the proliferation of hormone-independent CWR22Rv1 cells in culture (~ 5 µM) and also inhibits their growth in a prostate cancer xenograft murine model (10 mg/kg, i.p.daily).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
JMJD2C/KDM4C
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Jin, C., et al. 2014. Proc. Natl. Acad. Sci. USA111, 9235.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service