Skip to Content
Merck
All Photos(1)

Key Documents

SML2437

Sigma-Aldrich

H-151

H-151

≥98% (HPLC)

Synonym(s):

1-(4-Ethylphenyl)-3-(1H-indol-3-yl) urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.30
25 MG
€361.00

€89.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€89.30
25 MG
€361.00

About This Item

Empirical Formula (Hill Notation):
C17H17N3O
CAS Number:
Molecular Weight:
279.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€89.30


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1)NC(=O)Nc3ccc(cc3)CC)cccc2

InChI

1S/C17H17N3O/c1-2-12-7-9-13(10-8-12)19-17(21)20-16-11-18-15-6-4-3-5-14(15)16/h3-11,18H,2H2,1H3,(H2,19,20,21)

InChI key

UJZDIKVQFMCLBE-UHFFFAOYSA-N

Biochem/physiol Actions

Antagonist of the stimulator of interferon genes (STING) protein
H-151 is an irreversible antagonist of the stimulator of interferon genes (STING) protein, a central signalling component of the cytosolic DNA sensing pathway. Although STING is a vital component of the innate immune pathways, it has also been shown to have a pathogenic role in some inflammatory diseases. H-151 is a potent and selective STING antagonist with inhibitory activity both in human cells and in vivo in mice. Given as a pretreatment, H-151 was shown to attenuate pathological features of autoinflammatory disease in mouse models, reducing systemic cytokine responses in mice treated with the STING agonist 10-carboxymethyl-9-acridanone (CMA)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amir K Varkouhi et al.
Bioconjugate chemistry, 21(12), 2339-2346 (2010-11-06)
N,N,N-Trimethylated chitosan (TMC) is a biodegradable polymer emerging as a promising nonviral vector for nucleic acid and protein delivery. In the present study, we investigated whether the introduction of thiol groups in TMC enhances the extracellular stability of the complexes
J H Hamman et al.
Drug development and industrial pharmacy, 29(2), 161-172 (2003-03-22)
N-trimethyl chitosan chloride (TMC) is a polycation that enhances drug transport across epithelia by opening tight junctions. The degree of quaternization of TMC determines the number of positive charges available on the molecule for interactions with the negatively charged sites

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service