Skip to Content
Merck
All Photos(1)

Key Documents

5.04359

Sigma-Aldrich

Palmitoylethanolamide

Synonym(s):

Palmitoylethanolamide, N-(2-Hydroxyethyl)hexadecanamide, PEA, Palmidrol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€111.00

€111.00


Estimated to ship on27 May 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
€111.00

About This Item

Empirical Formula (Hill Notation):
C18H37NO2
CAS Number:
Molecular Weight:
299.49
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€111.00


Estimated to ship on27 May 2025Details


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 20 mM

storage temp.

−20°C

SMILES string

N(CCO)C(=O)CCCCCCCCCCCCCCC

InChI

1S/C18H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)19-16-17-20/h20H,2-17H2,1H3,(H,19,21)

InChI key

HXYVTAGFYLMHSO-UHFFFAOYSA-N

General description

An endogenous lipid that acts as a selective agonist for GPR55 receptors (EC50 = 4, 19 800 and >30 000 nM at GPR55, CB2 and CB1 receptors respectively). Producing robust anti-inflammatory actions by activating PPARα (EC50 = 3 µM).

Biochem/physiol Actions

Primary Target
GPR55 receptor

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming is required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Griffin, G., et al. 2000. J. Pharmacol. Exp. Ther.292, 886.

Lambert, D., et al. 2001. Epilepsia.42, 321.
Lambert, D., et al. 2002. Curr. Med. Chem.9, 663.
Lo Verme, J., et al. 2005. Life Sci.77, 1685.
Re, G., et al. 2005. Vet. J.173, 21.
Ryberg, R., et al. 2007. Br. J. Pharmacol.152, 1092.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service