Skip to Content
Merck
All Photos(1)

Key Documents

495417-M

Millipore

N-Oleoyl Ethanolamine

A potent inhibitor of the ceramidase found in human kidney and cerebellum.

Synonym(s):

N-Oleoyl Ethanolamine, N-2-Hydroxyethyl-9(Z)-octadecenamide, AC Inhibitor I, Acid Ceramidase Inhibitor I, OEA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H39NO2
CAS Number:
Molecular Weight:
325.53
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

chloroform: 5 mg/mL
ethanol: 5 mg/mL
methanol: 5 mg/mL

storage temp.

−20°C

SMILES string

N(CCO)C(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C20H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)21-18-19-22/h9-10,22H,2-8,11-19H2,1H3,(H,21,23)/b10-9-

InChI key

BOWVQLFMWHZBEF-KTKRTIGZSA-N

General description

A potent inhibitor of the ceramidase found in human kidney and cerebellum. Blocks the conversion of ceramide to sphingosine. A specific inhibitor of acid ceramidase (IC50 ~ 500 µM).

Biochem/physiol Actions

Primary Target
Acid ceramidase
Target IC50: ~ 500 µM against acid ceramidase

Warning

Toxicity: Standard Handling (A)

Other Notes

Pahan, K., et al. 1998. J. Biol. Chem. 273, 2591.
Ping, S.E., and Barrett, G.L. 1998. J. Neurosci. Res. 54, 206.
Sugita, M., et al. 1975. Biochim. Biophys. Acta398, 125.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service