Skip to Content
Merck
All Photos(1)

Key Documents

492027-M

Sigma-Aldrich

NRG1 Isoform SMDF, Human, Recombinant

Synonym(s):

Heregulin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

Assay

>95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0 EU/μg protein Endotoxin

storage temp.

−20°C

General description

Recombinant, human neuregulin 1 (NRG1), isoform SMDF (sensory and motor neuron-derived factor), consisting of amino acids 1-263, 117-296, and 162-296, each fused at the C-terminus to a His•Tag sequence and expressed in S. frugiperda Sf21 cells using a baculovirus expression system. The neuregulin (also known as heregulin) family of cytokines is comprised of multiple secreted or membrane-bound isorforms that are produced from the single heregulin gene by alternative splicing and/or usage of different promoters. NRG1 isoform SMDF is a type III isoform with a C-terminal EGF-like domain that interacts with the erbB family of tyrosine kinase receptors. NRG1 isoform SMDF expression has been found to be restricted to the nervous system and is believed to play an important role in the development of the nervous system.

Biochem/physiol Actions

ED₅₀ = 1-5 ng/ml as measured in a serum-free proliferation assay using MCF-7 human breast adenocarcinoma cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filtered PBS, BSA.

Reconstitution

Following reconstituion refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 100 µg/ml with sterile PBS containing ≥1% human or bovine serum albumin.

Other Notes

Meyer, D. et al. 1997. Development124, 3575.
Yarden, Y. and D. Wen 1994. in Guidebook to Cytokinesand Their Receptors, N.A. Nicola, Ed., Oxford University Press, p. 146.
Karey, K.P., et al. 1988. Cancer Res.48, 4083.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service