Skip to Content
Merck
All Photos(1)

Key Documents

492028

Sigma-Aldrich

NRG1-β1/HRG1-β1 ECD, Human, Recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€619.00

€619.00


Estimated to ship on10 April 2025Details



Select a Size

Change View
50 μG
€619.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.77

€619.00


Estimated to ship on10 April 2025Details


Assay

≥90% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, human neuregulin 1-β1/heregulin 1-β1 extracellular domain (NRG1-beta1/HRG1-beta1 ECD) consisting of amino acids 2-246 and expressed in E. coli. All NRG1 isoforms contain an EGF-like domain (β-splice variant) that is used to directly bind to the ErbB3 or ErbB4 receptor tyrosine kinases.

Biochem/physiol Actions

ED₅₀ = 2.5-12.5 ng/ml as measured in a serum-free proliferation assay using MCF7 human breast adenocarcinoma cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filtered acetonitrile, TFA, BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short term-storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute 50 100 µg/ml with sterile PBS containing ≥1% human or bovine serum albumin.

Other Notes

Buonanno, A., and G.D Fischbach 2001. Curr. Opin. Neurobiol.11, 287.
Adlkofer, K. and C. Lai 2000. Glia29, 104.
Garratt, A.N. et al., 2000. BioEssays22, 987.
Karey, K.P., et al. 1988. Cancer Res.48, 4083.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service