Skip to Content
Merck

4.80939

SAFC

Phosphoric acid 1 mol/L

EMPROVE® EXPERT

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€29.00
950 L
€6,309.00

€29.00


Estimated to ship on02 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 L
€29.00
950 L
€6,309.00

About This Item

UNSPSC Code:
12352323

€29.00


Estimated to ship on02 April 2025Details


Request a Bulk OrderRequest a Sample

product line

EMPROVE® EXPERT

Quality Level

form

liquid

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service