Skip to Content
Merck
All Photos(1)

Key Documents

P8192

Sigma-Aldrich

TWEEN® 80 solution

10%, low peroxide

Synonym(s):

Polysorbate 80, Polyethylene glycol sorbitan monooleate solution, Polyoxyethylenesorbitan Monooleate

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€40.80

€40.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€40.80

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

€40.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

description

non-ionic

concentration

10%

storage temp.

2-8°C

SMILES string

O1C(CC(C1C(OCCO)COCCOC(=O)CCCCCCC\C=C\CCCCCCCC)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic detergent used for selective protein extraction and isolation of nuclei from mammalian cell lines

Components

Contains BHT as an antioxidant.

Quality

Low Peroxide

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R G Deshpande et al.
Journal of medical microbiology, 46(3), 233-238 (1997-03-01)
Contact-dependent haemolytic activity was observed with cells of Mycobacterium tuberculosis H(37)Rv and M. tuberculosis H(37)Ra, but not with those of M. bovis, M. bovis BCG and M. africanum. Culture filtrates of all these strains did not exhibit any haemolytic activity.
Isolation of subcellular organelles.
B Storrie et al.
Methods in enzymology, 182, 203-225 (1990-01-01)
Mylène Tajan et al.
Nature communications, 12(1), 366-366 (2021-01-16)
Many tumour cells show dependence on exogenous serine and dietary serine and glycine starvation can inhibit the growth of these cancers and extend survival in mice. However, numerous mechanisms promote resistance to this therapeutic approach, including enhanced expression of the de

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service