Skip to Content
Merck
All Photos(1)

Key Documents

444256

Sigma-Aldrich

MMP Substrate III, Fluorogenic

A specific fluorogenic resonance energy transfer (FRET) peptide substrate with an MMP cleavable Gly-Leu bond and EDANS/Dabcyl as fluorophore/quencher combination.

Synonym(s):

MMP Substrate III, Fluorogenic, DABCYL-GABA-PQGL-E(EDANS)-AK-NH₂, TNO211

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€343.00

€343.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 μG
€343.00

About This Item

Empirical Formula (Hill Notation):
C63H88N16O14S
Molecular Weight:
1325.54
UNSPSC Code:
12352200
NACRES:
NA.77

€343.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

DMSO: soluble
Tris-buffered saline, pH 7.6: soluble

fluorescence

λex ~340 nm
λem ~485 nm

shipped in

ambient

storage temp.

−20°C

General description

A fluorescence energy transfer (FRET) peptide substrate with MMP cleavable Gly-Leu bond and EDANS/Dabcyl as fluorophore/quencher combination. Useful for the detection of MMP activity (kcat/Km = 619,000 M-1s-1 for MMP-2, 206,000 M-1s-1 for MMP-9, 40,000 M-1s-1 for MMP-3, 21,000 M-1s-1 for MMP-1; pH = 7.6 at 37°C). Exhibits a high degree of sensitivity, which is not affected by optical disturbances in biological media. Excitation max.: ~340 nm; emission max.: ~485 nm.
A specific fluorogenic resonance energy transfer (FRET) peptide substrate with an MMP cleavable Gly-Leu bond and EDANS/Dabcyl as fluorophore/quencher combination. Useful for the detection of MMP activity [kcat/Km = 619,000 M-1s-1 for MMP-2, 206,000 M-1s-1 for MMP-9, 40,000 M-1s-1 for MMP-3, and 21,000 M-1s-1 for MMP-1; at 37°C, pH 7.6 ]. Exhibits a high degree of sensitivity that is not affected by optical disturbances in biological media. Also useful for MMP activity measurements in synovial fluid and culture medium.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Detection of MMP activity
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

DABCYL-GABA-Pro-Gln-Gly-Leu-Glu(EDANS)-Ala-Lys-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Beekman, B., et al. 1996. FEBS Lett390, 221.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service