Skip to Content
Merck
All Photos(1)

Key Documents

03-32-5032

Sigma-Aldrich

MMP-2/MMP-7 Substrate, Fluorogenic

Extremely sensitive fluorogenic substrate for matrix metalloproteinases (MMPs).

Synonym(s):

MMP-2/MMP-7 Substrate, Fluorogenic

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€270.00

€270.00


Estimated to ship on28 April 2025Details



Select a Size

Change View
1 MG
€270.00

About This Item

Empirical Formula (Hill Notation):
C49H68N14O15
Molecular Weight:
1093.15
UNSPSC Code:
12352209
NACRES:
NA.83

€270.00


Estimated to ship on28 April 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

yellow

solubility

20% acetic acid: 1 mg/mL

fluorescence

λex ~325 nm
λem ~393 nm

shipped in

ambient

storage temp.

−20°C

General description

Extremely sensitive fluorogenic substrate for matrix metalloproteinases (MMPs). Suitable for assessment of total activity in crude preparations by continuous assays (kcat/Km = 1.7 x 105 M-1sec-1 for MMP-7 and 6.3 x 105 M-1sec-1 for MMP-2).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Fluorogenic substrate for matrix metalloproteinases (MMPs)
Product does not compete with ATP.
Reversible: no
kcat/Km = 1.7 x 105 M-1sec-1 for MMP-7 and 6.3 x 105 M⁻¹ sec⁻¹ for MMP-2

Warning

Toxicity: Standard Handling (A)

Sequence

MCA-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH₂ ·TFA [Dpa = N-3-(2,4-dinitrophenyl)-L-2,3-diaminopropionyl]

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Knight, C.G., et al. 1992. FEBS Lett. 296, 263.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service