Skip to Content
Merck
All Photos(1)

Key Documents

435300

Sigma-Aldrich

LFM-A13

A potent, cell-permeable, reversible, substrate competitive, and specific inhibitor of Bruton′s Tyrosine Kinase (BTK; IC₅₀ = 17.2 µM for human BTK in vitro and IC₅₀ = 2.5 µM for recombinant BTK).

Synonym(s):

LFM-A13, α-Cyano-β-hydroxy-β-methyl-N-(2,5-dibromophenyl)propenamide, BTK Inhibitor I, Bruton′s Tyrosine Kinase Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€163.00

€163.00


Estimated to ship on24 April 2025Details



Select a Size

Change View
5 MG
€163.00

About This Item

Empirical Formula (Hill Notation):
C11H8Br2N2O2
CAS Number:
Molecular Weight:
360.00
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€163.00


Estimated to ship on24 April 2025Details


Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light brown

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C11H8Br2N2O2/c1-6(16)8(5-14)11(17)15-10-4-7(12)2-3-9(10)13/h2-4,16H,1H3,(H,15,17)

InChI key

UVSVTDVJQAJIFG-UHFFFAOYSA-N

General description

A potent, cell-permeable, reversible, substrate competitive, and specific inhibitor of Bruton’s Tyrosine Kinase (BTK; IC50 = 17.2 µM for human BTK in vitro and IC50 = 2.5 µM for recombinant BTK). Also inhibits Polo-like kinase in an ATP-competitive manner (IC50 = 10 µM and 61 µM for Plx1 and Plk3, respectively), and displays antitumor properties. Does not affect the enzymatic activity of other protein tyrosine kinases, including EGFR, HCK, IRK JAK1, and JAK3 even at concentrations of 278 µM. Enhances the chemosensitivity of BTK-positive B-lineage leukemia cells to ceramide- and vincristine-induced apoptosis.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Human Bruton′s Tyrosine Kinase (BTK)
Product competes with ATP.
Reversible: yes
Target IC50: 17.2 µM for human BTK in vitro and 2.5 µM for recombinant BTK

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Uckun, F.M., et al. 2006. Bioorg. Med. Chem. In press.
Mahajan, S., et al. 1999. J. Biol. Chem. 274, 9587.
Vassilev, A., et al. 1999. J. Biol. Chem. 274, 1646.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service