Skip to Content
Merck
All Photos(1)

Key Documents

G211280

Sigma-Aldrich

DMT-2′Fluoro-dG(ib) Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym(s):

DMT-2′Fluoro-dG(ib) Amidite

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€168.00
2 G
€321.00

€168.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€168.00
2 G
€321.00

About This Item

Empirical Formula (Hill Notation):
C44H53FN7O8P
CAS Number:
Molecular Weight:
857.91
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.21

€168.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

impurities

≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for PerkinElmer
configured for Polygen

nucleoside profile

base: deoxyadenosine
base protecting group: benzoyl
2' protecting group: fluoro
5' protecting group: DMT
deprotection: standard

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](F)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)C(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C44H53FN7O8P/c1-27(2)40(53)49-43-48-39-37(41(54)50-43)47-26-51(39)42-36(45)38(60-61(58-24-12-23-46)52(28(3)4)29(5)6)35(59-42)25-57-44(30-13-10-9-11-14-30,31-15-19-33(55-7)20-16-31)32-17-21-34(56-8)22-18-32/h9-11,13-22,26-29,35-36,38,42H,12,24-25H2,1-8H3,(H2,48,49,50,53,54)/t35-,36-,38-,42-,61?/m1/s1

InChI key

KJFUMXZZVYMQEU-AOCJBPQJSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service