Skip to Content
Merck
All Photos(1)

Key Documents

3801-M

Sigma-Aldrich

Saffron

Harleco®, suitable for Gram staining

Synonym(s):

Saffron, Gram Stain, C.I. 75100

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
976.96
MDL number:
UNSPSC Code:
41116121
EC Index Number:
255-881-6

form

powder

Quality Level

manufacturer/tradename

Harleco®

technique(s)

Gram staining: suitable

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)OC[C@H]2O[C@H]([C@@H]([C@H]([C@@H]2O)O)O)OC(=O)\C(=C\C=C\C(=C\C=C\C=C(/C)\C=C\C=C(/C)\C(=O)O[C@@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)CO)\C)\C

InChI

1S/C44H64O24/c1-19(11-7-13-21(3)39(59)67-43-37(57)33(53)29(49)25(65-43)17-61-41-35(55)31(51)27(47)23(15-45)63-41)9-5-6-10-20(2)12-8-14-22(4)40(60)68-44-38(58)34(54)30(50)26(66-44)18-62-42-36(56)32(52)28(48)24(16-46)64-42/h5-14,23-38,41-58H,15-18H2,1-4H3/b6-5+,11-7+,12-8+,19-9+,20-10+,21-13+,22-14+/t23-,24-,25-,26-,27-,28-,29-,30-,31+,32+,33+,34+,35-,36-,37-,38-,41-,42-,43+,44+/m1/s1

InChI key

SEBIKDIMAPSUBY-RTJKDTQDSA-N

General description

HARLECO® For gram staining

Application

For In-Vitro Diagnostic Use.
For Professional Use.
HARLECO® For gram staining

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service