Skip to Content
Merck
All Photos(1)

Key Documents

1.11662

Millipore

CHAPS

for biochemistry

Synonym(s):

CHAPS, 3-[(3-Cholamidopropyl)dimethylammonio] propane sulfonate), 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€336.00

€336.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
€336.00

About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S
CAS Number:
Molecular Weight:
614.88
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.05

€336.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Product Name

CHAPS (3-[(3-Cholamidopropyl)-dimethylammonio]-propane- sulfonate), for biochemistry

Quality Level

form

solid

mol wt

micellar avg mol wt 6150

aggregation number

10

pH

6 (100 g/L in H2O)

CMC

6 mM (20-25°C)

mp

157 °C

transition temp

cloud point >100 °C

storage temp.

2-8°C

SMILES string

C[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)CC4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C32H58N2O7S/c1-21(8-11-29(38)33-14-6-15-34(4,5)16-7-17-42(39,40)41)24-9-10-25-30-26(20-28(37)32(24,25)3)31(2)13-12-23(35)18-22(31)19-27(30)36/h21-28,30,35-37H,6-20H2,1-5H3,(H-,33,38,39,40,41)/t21-,22?,23-,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

UMCMPZBLKLEWAF-RFCNGIAKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

CHAPS is a nondenaturing zwitterionic detergent for membrane biochemistry.
Useful for solubilizing membrane proteins and breaking protein-protein interactions. CHAPS′ small micellar molecular weight (6,150) and high critical micelle concentration (6-10 mM) allow it to be removed from samples by dialysis. It is also suitable for protein solubilization for isoelectric focusing and two-dimensional electrophoresis. CHAPS is commonly used for non-denaturing (without urea) IEF and has been shown to give excellent resolution of some subcellular preparations and plant proteins. Concentrations between 2-4% (w/v) are typically used in an IEF gel.

Analysis Note

Assay (ex N): ≥ 99 %
Identity (IR-spectrum): passes test
Absorbance A (280 nm; 1 %; 1 cm; water): ≤ 0.1
Absorbance A (260 nm; 1 %; 1 cm; water): ≤ 0.1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service