Skip to Content
Merck
All Photos(1)

Documents

266788

Sigma-Aldrich

Diacylglycerol Kinase Inhibitor II

The Diacylglycerol Kinase Inhibitor II, also referenced under CAS 120166-69-0, controls the biological activity of Diacylglycerol KInase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Diacylglycerol Kinase Inhibitor II, R59949, 3-{2-[4-[ bis-(4-Fluorophenyl)methylene]-1-piperidinyl]ethyl}-2,3-dihydro-2-thioxo-4(1H)quinazolinone, DGK Inhibitor II, R59949, 3-{2-[4-[bis-(4-Fluorophenyl)methylene]-1-piperidinyl]ethyl}-2,3-dihydro-2-thioxo-4(1H)quinazolinone, DGK Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H25F2N3OS
CAS Number:
Molecular Weight:
489.58
MDL number:
UNSPSC Code:
41121800
NACRES:
NA.54

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 12.5 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C28H25F2N3OS/c29-22-9-5-19(6-10-22)26(20-7-11-23(30)12-8-20)21-13-15-32(16-14-21)17-18-33-27(34)24-3-1-2-4-25(24)31-28(33)35/h1-12H,13-18H2,(H,31,35)

InChI key

ZCNBZFRECRPCKU-UHFFFAOYSA-N

General description

Inhibitor useful for elucidating roles of protein kinase C. Inhibits diacylglycerol (DAG) kinase in isolated platelet membranes (IC50 = 300 nM) and in intact platelets (IC50 = 120 nM). A more potent analog of DAG kinase inhibitor I.
Inhibits diacylglycerol kinase in isolated platelet membranes (IC50 = 300 nM) and in intact platelets (IC50 = 120 nM) by binding to the catalytic domain. A more potent analog of diacylglycerol kinase inhibitor I (Cat. No. 266785).

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no
Target IC50: 300 nM and 120 nM against diacylglycerol kinase in isolated platelet membranes and in intact platelets, respectively

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot, flush with an inert gas and freeze at -20°C. Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Jiang, Y., et al. 2000. Biochem. Pharmacol. 59, 763.
Jones, D.R., et al. 1999. J. Biol. Chem.274, 16846.
Rodriguez-Linares, B., et al. 1991. Biochem. Pharmacol. 41, 835.
de Chaffoy de Courcelles, D., et al. 1989. J. Biol. Chem.264, 3274.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan V Outram et al.
Immunology, 105(4), 391-398 (2002-05-03)
The diacylglycerol kinases (DGK) form a family of isoenzymes that catalyse the conversion of diacylglycerol (DAG) to phosphatidic acid (PA), both powerful second messengers in the cell. DGKalpha is expressed in brain, peripheral T cells and thymocytes and has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service