Skip to Content
Merck
All Photos(1)

Documents

DUO84012

Sigma-Aldrich

Duolink® flowPLA Compensation Beads - Green

For flow cytometry standard and multiplex analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.32

Quality Level

product line

Duolink®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
multiplexing: suitable
proximity ligation assay: suitable

fluorescence

λex 498 nm; λem 523 nm (excitation laser line 488 nm)

suitability

suitable for fluorescence

shipped in

wet ice

storage temp.

2-8°C

Application

Duolink® proximity ligation assay(PLA®) allows for endogenous detection of protein interactions, post translational modifications, and protein expression levels at the single molecule level in fixed cells and tissue samples.

It is recommended that Duolink® compensation beads should be used when performing a multiplexed experiment (DUO96001) to save on time and reagent cost, rather than using precious sample. Please note that these Green beads are specific to the Green fluor utilized in the Duolink® Multicolor FlowPLA kit, not the Green fluorophore in the stand-alone Duolink® microscopy and flow kits. This product can be used to determine flow cytometry instrumentation collection parameters, inform experimental design, and as a control for compensation data for multiplexed FlowPLA experiments.

Visit our Duolink® PLA Resource Center for information on how to run a Duolink® experiment, applications, troubleshooting, and more.

Duolink® Multicolor flow PLA Reagent Pack Kits used with Duolink® PLA Multicolor Probemaker Kits will enable Multiplexing of up to 4 protein events with sensitive detection of proteins, protein-protein interactions, and protein modifications within cell populations by flow cytometry. To perform a Duolink® flowPLA experiment, you will need fixed and permeabilized suspended cells, and at least two primary antibodies that specifically recognize your proteins of interest. Analysis is carried out using standard flow cytometry assay equipment. User must provide a fixed cell suspension and primary antibodies.
Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC) or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Duolink® in situ reagents are suitable for use on fixed cells, cytospin cells, cells grown on slide, formalin-fixed, paraffin embedded (FFPE), or tissue (fresh or frozen). Flow validated antibodies are recommended.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

View full Duolink® product list

Features and Benefits

  • Save time and reagent cost by utilizing beads for compensation rather than precious sample and multiplex reagents
  • No overexpression or genetic manipulation required
  • High specificity (fewer false positives)
  • Single molecule sensitivity due to rolling circle amplification
  • Relative quantification possible
  • No special equipment needed
  • Quicker and simpler than FRET
  • Increased accuracy compared to co-IP
  • Publication-ready results

Components

This product is comprised of the following:
6.0 - 8.0μm polystyrene beads suitable for use in flow cytometry assays. The bead slurry should have a concentration of 1.5-3.0 million beads/mL. Green beads can be mixed with the negative control compensation (DUO84010), or collected alone.

Legal Information

Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service