Skip to Content
Merck
All Photos(2)

Key Documents

218860

Sigma-Aldrich

Casein Kinase II Inhibitor VIII

The Casein Kinase II Inhibitor VIII controls the biological activity of Casein Kinase II. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Casein Kinase II Inhibitor VIII, CK2 Inhibitor VIII, DYRK1B Inhibitor, Flt-3 Inhibitor VII, 4-(2-(4-Methoxybenzamido)thiazol-5-yl)benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€205.00

€205.00


Estimated to ship on17 April 2025Details



Select a Size

Change View
10 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C18H14N2O4S
CAS Number:
Molecular Weight:
354.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on17 April 2025Details


Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(OC)C=C1)NC2=NC=C(C3=CC=C(C(O)=O)C=C3)S2

InChI

1S/C18H14N2O4S/c1-24-14-8-6-12(7-9-14)16(21)20-18-19-10-15(25-18)11-2-4-13(5-3-11)17(22)23/h2-10H,1H3,(H,22,23)(H,19,20,21)

InChI key

UPIVZLFKYMNZDG-UHFFFAOYSA-N

General description

A cell-permeable thiazolamide compound that acts as a potent, reversible, and ATP-competitive inhibitor against casein kinase-2/CK2 (IC50 = 32 and 46 nM against CK2α and CK2α′, respectively), as well as DYRK1B and FLt-3 (82% and 76% inhibition, respectively, at 30 nM), while diplaying reduced or little potency toward other targets in a 70-kinase panel selectivity profile study. Shown to effectively block the proliferation of a variety of cancer cells (IC50 = 2.6, 1.6, and 2.4 µM, in A549, HCT116, and MCF-7 cultures, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CK2
Reversible: yes
Secondary Target
DYRK1B, Flt-3
Target IC50: 32 and 46 nM for CK2&alpha

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hou, Z., et al. 2012. J. Med. Chem.55, 2899.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service