Skip to Content
Merck
All Photos(1)

Key Documents

218710

Sigma-Aldrich

Casein Kinase II Inhibitor III, TBCA

The Casein Kinase II Inhibitor III, TBCA, also referenced under CAS 934358-00-6, controls the biological activity of Casein Kinase II. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Casein Kinase II Inhibitor III, TBCA, (E)-3-(2,3,4,5-Tetrabromophenyl)acrylic acid, CKII Inhibitor, TBCA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€176.00

€176.00


Estimated to ship on14 April 2025Details



Select a Size

Change View
5 MG
€176.00

About This Item

Empirical Formula (Hill Notation):
C9H4Br4O2
CAS Number:
Molecular Weight:
463.74
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

€176.00


Estimated to ship on14 April 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

potency

110 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL (Use only fresh DMSO.)
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Brc1c(c(cc(c1Br)\C=C\C(=O)O)Br)Br

InChI

1S/C9H4Br4O2/c10-5-3-4(1-2-6(14)15)7(11)9(13)8(5)12/h1-3H,(H,14,15)/b2-1+

InChI key

SVJQCVOKYJWUBC-OWOJBTEDSA-N

General description

A cell-permeable tetrabrominated cinnamic acid compound that acts as a potent and ATP-competitive inhibitor of Casein Kinase II (IC50 = 110 nM, Ki = 77 nM) with greater selectivity than TBB (Cat. Nos. 218697 and 218708) against a panel of 28 commonly studied kinases. Shown to efficiently induces apoptosis in Jurkat cells (DC50 = 7.7 µM). Also available as a 25 mM solution in DMSO (Cat. No. 218861).

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: no
Target Ki: 77 nM against Casein Kinase II

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Pagano, M.A., et al. 2006. Chembiochem8, 129

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service