Skip to Content
Merck
All Photos(1)

Key Documents

2102-M

Millipore

Protein-Concentrate Kit (Med)

Sign Into View Organizational & Contract Pricing

Select a Size

2102-M
€276.00

€276.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
2102-M
€276.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.27

€276.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

application(s)

sample preparation

General description

The Chemicon Protein-Concentrate kit uses a proprietary reagent Universal Protein Precipitation Agent (UPPA). Protein solutions as dilute as 1 ng/mL can be quantitatively concentrated into a small volume. Protein precipitation is not effected by the presence of detergents, chaotropic, or other common laboratory agents. After precipitation, the precipitate is suspended in a small volume of Precipitate Suspension Buffer supplied with each kit. If the protocol is followed correctly, the recovery is generally 100%. There are enough reagents for concentrating up to 30 mL of dilute protein solution.

Application

Suitable for concentrating proteins for running gels, raising antibodies, protein purification, protein assays and other applications.

This kit is not suitable for those proteins which may lose some of their biological activities when precipitated.

PROTOCOL: ImportantPerform the entire procedure in the cold (ice bucket) unless specified otherwise. Concentration should be performed in a centrifuge tube. For small volumes, use microfuge tubes. Always position microfuge tubes in the centrifuge at the same orientation, i.e. cap-hinge facing out-ward, this will allow the pellet to remain glued to the same side of the tube during repeated centrifugations and minimize the loss of protein pellets. See product insert for protocol.

Packaging

1 kit

Components

UPPA-I 100 mL UPPA-II 100 mL OrgoSol 2 x 50 mL UPC-SEED 2 x 0.30 mL Precipitate Buffer I 2 x 2.0 mL Precipitate Buffer II 2 x 0.5 mL UCP-Wash 2 x 2.0 mL

Storage and Stability

Store at room temperature for up to 6 months from date of purchase. To avoid possible microbial contamination, dispense all solutions aseptically.

Legal Information

UCP is a trademark of Intelligent Material Solutions, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

-2.2 °F

Flash Point(C)

-19 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service