Skip to Content
Merck
All Photos(1)

Key Documents

204896

Sigma-Aldrich

Complement C8, Human

Complement C8, Human, CAS 80295-58-5, is a native glycoprotein composed of three non-identical subunits of 64, (α), 64 (β), and 22 kDa (γ).

Synonym(s):

Complement C8, Human

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€541.00

€541.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
€541.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

€541.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥85% (SDS-PAGE)

form

liquid

specific activity

≥150,000 C8H50 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

IgG, IgA, IgM, albumin, C3, C4, C7, C9, trace

shipped in

dry ice

storage temp.

−70°C

General description

Native, human C8 complement component. Glycoprotein composed of three non-identical subunits of M.W. 64 kDa (α), 64 kDa (β), and 22 kDa (γ). Present in normal human serum at 55 µg/ml. On activation of complement via either the classical or alternative pathway, C5 is cleaved into C5a and C5b fragments. The nascent C5b fragment binds to C6, C7, and C8 resulting in formation of the C5b-8 complex on target surfaces. Each membrane-bound, hydrophobic C5b-8 complex combines with three to six C9 molecules to complete the assembly of the lytic C5b-9 complement membrane attack complex (MAC).

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.2.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Steckel, E.W., et al. 1980. J. Biol. Chem. 255, 11997.
Petersen, B.H., et al. 1976. J. Clin. Invest. 57, 283.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service