Skip to Content
Merck
All Photos(1)

Documents

204001

Sigma-Aldrich

BX-795

BX-795, also referenced under CAS 702675-74-9, modulates the biological activity of multiple kinases. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

BX-795, AMPK Related Kinase 5 Inhibitor I, ARK5 Inhibitor I, NUAK Kinase Inhibitor I, NUAK1 Inhibitor I, SnF1 Like Kinase-1 Inhibitor I, N-(3-((5-Iodo-4-((3-(2-thienylcarbonyl)amino)propyl)amino)-2-pyrimidinyl)amino)phenyl)-1-pyrrolidinecarboxamide, Aurora Kinase, N-(3-((5-Iodo-4-((3-(2-thienylcarbonyl)amino)propyl)amino)-2-pyrimidinyl)amino)phenyl)-1-pyrrolidinecarboxamide, AMPK Related Kinase 5 Inhibitor I, ARK5 Inhibitor I, NUAK Kinase Inhibitor I, NUAK1 Inhibitor I, SnF1 Like Kinase-1 Inhibitor I, Aurora Kinase Inhibitor VIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26IN7O2S
CAS Number:
Molecular Weight:
591.47
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C23H26IN7O2S/c24-18-15-27-22(30-20(18)25-9-5-10-26-21(32)19-8-4-13-34-19)28-16-6-3-7-17(14-16)29-23(33)31-11-1-2-12-31/h3-4,6-8,13-15H,1-2,5,9-12H2,(H,26,32)(H,29,33)(H2,25,27,28,30)

InChI key

VAVXGGRQQJZYBL-UHFFFAOYSA-N

General description

A cell-permeable aminopyrimidine compound that acts as a potent, ATP-competitive and reversible multi-kinase inhibitor (IC50 = 11 nM for PDK1 at ≤ 10 µM of ATP; IC50 = 5 , 6, 19, 31, 41 and 111 nM for NUAK1, TBK1, MARK4, Aurora B, IKKε and PDK1 at 100 µM of ATP, respectively) and displays 50-fold greater selectivity over Cdk2/E, EGFR, GSK-3β, IR, c-Kit, PKA, PKC, TAK1 and VEGFR. Shown to block PDK1/Akt signaling, induce growth arrest and apoptosis in PC-3 and MDA-468 cells (IC50 = 0.25 and 0.75 µM, respectively); further, inhibit phosphorylation (pSer396), nuclear translocation and transcriptional activity of IRF3 and reduce IFN-β production in stimulated macrophages.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Clark, K., et al. 2009. J. Biol. Chem.284, 14136.
Tamgumcy, T., et al. 2008. Exp. Cell Res.314, 2299.
Feldman, R.I., et al. 2005. J. Biol. Chem.280, 19867.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah A Scuderi et al.
Oncology research, 28(7), 779-790 (2021-03-21)
Glioma are common malignant brain tumors, among which glioblastoma multiforme (GBM) has the worst prognosis. Different studies of GBM revealed that targeting nuclear factor B (NF-B) induced an attenuation tumor proliferation and prolonged cell survival. TBK1 {TANK [TRAF (TNF (tumor-necrosis-factor)
Lianxin Hu et al.
Cancer discovery, 10(3), 460-475 (2019-12-08)
TANK binding kinase 1 (TBK1) is an important kinase involved in the innate immune response. Here we discover that TBK1 is hyperactivated by von Hippel-Lindau (VHL) loss or hypoxia in cancer cells. Tumors from patients with kidney cancer with VHL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service