Skip to Content
Merck
All Photos(1)

Key Documents

20-159

Sigma-Aldrich

5M NaCl - for use in Chromatin Immunoprecipitation (ChIP)

5M NaCl -for use in Chromatin Immunoprecipitation (ChIP).

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
€182.00

€182.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 μL
€182.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

€182.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Quality Level

form

liquid

manufacturer/tradename

Upstate®

shipped in

wet ice

Application

For use in Chromatin Immunoprecipitation assays.
Research Category
All

Physical form

5M NaCl

Storage and Stability

1 year at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutual interaction and reciprocal down-regulation between c-met and insulin receptor substrate-1.
Tiziana DeAngelis,Andrea Morrione,Renato Baserga
Journal of Cellular Physiology null
Insulin receptor substrate-1 regulates the transformed phenotype of BT-20 human mammary cancer cells.
Ozlem Dalmizrak,An Wu,Jia Chen,Hongzhi Sun,Fransiscus E Utama,Diana Zambelli,Thai H Tran et al.
Cancer Research null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service