Skip to Content
Merck
All Photos(1)

Key Documents

194434

Sigma-Aldrich

ABTS Chromophore

Synonym(s):

ABTS Chromophore

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€167.00

€167.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€167.00

About This Item

Empirical Formula (Hill Notation):
C18H18N4O6S4
Molecular Weight:
514.62
UNSPSC Code:
41116133
NACRES:
NA.41

€167.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

form

solution

Quality Level

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clearvery pale yellow

shipped in

ambient

storage temp.

2-8°C

General description

Peroxidase substrate that is soluble, safe, convenient and has good overall performance in ELISA. ABTS Chromophore develops intense blue-green color that can be measured at 405 nm to 410 nm. May be used for both kinetic and endpoint reactions.
Single component peroxidase substrate which is soluble, safe, convenient and has good overall performance for ELISA. In the presence of horseradish peroxidase (HRP) and hydrogen peroxide, ABTS is oxidized to an intense blue-green, water-soluble product. This product is a radical cation having an absorption maximum at 416 nm. Typically, product formation is measured spectrophotometrically at 405 nm, allowing a quantitative analysis of HRP-based systems. The color formation as a function of time may be monitored, or the reaction may be stopped by the addition of acid; since the acid does not alter the absorbance at 405 nm, one may perform both kinetic and endpoint measurements at the same wavelength.

Warning

Toxicity: Irritant (B)

Physical form

Contains optimal concentrations of ABTS and H₂O₂ in a citrate buffer (pH 4.0) with proprietary stabilizers.

Reconstitution

ABTS solution should be stored at 4°C, but must be warmed to room temperature before use. The solution should be protected from exposure to direct sunlight, and should be discarded if it becomes bright yellow or turbid.

Other Notes

Rodgriquez-Lopez, J.N., et al. 2000. Biochemistry39, 13201.

Legal Information

ABTS is a trademark of Boehringer Mannheim Corp.
ABTS is a trademark of Roche
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service