Skip to Content
Merck
All Photos(1)

Key Documents

17-128

Sigma-Aldrich

Alkaline/Acid Phosphatase Assay Kit (R-R-A-pS-V-A)

Alkaline/Acid Phosphatase Assay Kit is routinely used to detect phosphatase activity by either dephosphorylation of the phosphopeptide (RRApSVA) or hydrolysis of pNPP.

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€704.00

€704.00


Estimated to ship on02 May 2025Details



Select a Size

Change View
1 KIT
€704.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

€704.00


Estimated to ship on02 May 2025Details


Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

detection method

colorimetric

shipped in

wet ice

Application

Used to detect/quantify: Alkaline/Acid

Packaging

Kit capacity: 100 assays

Components

Malachite Green Solution A (Cat.# 20-105)

Malachite Green Additive (Cat.# 20-104)

Phosphate Standard (Cat.# 20-103)

Serine Phosphopeptide (RRApSVA) (Cat.# 12-220)

pNPP (p-Nitrophenyl Phosphate) (Cat.# 20-106)

NiCl2, 40mM (Cat.# 20-178)

pNPP Ser/Thr Assay Buffer (Cat.# 20-179)

96-well microtiter plate

Quality

routinely used to detect phosphatase activity by either dephosphorylation of the phosphopeptide (RRApSVA) or hydrolysis of pNPP

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1A Inhalation - Met. Corr. 1 - Repr. 1B - Skin Sens. 1 - STOT RE 2

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of subnanomolar amounts of phosphate bound to seryl and threonyl residues in phosphoproteins using alkaline hydrolysis and malachite green.
Ekman, P and Jager, O
Analytical biochemistry, 214, 138-141 (1993)
An investigation of the substrate specificity of protein phosphatase 2C using synthetic peptide substrates; comparison with protein phosphatase 2A
Donella Deana, A., et al
Biochimica et Biophysica Acta, 1051, 199-202 (1990)
Nathan J Castro et al.
Tissue engineering. Part A, 22(13-14), 940-948 (2016-06-15)
Osseous tissue defects caused by trauma present a common clinical problem. Although traditional clinical procedures have been successfully employed, several limitations persist with regards to insufficient donor tissue, disease transmission, and inadequate host-implant integration. Therefore, this work aims to address
P P Van Veldhoven et al.
Analytical biochemistry, 161(1), 45-48 (1987-02-15)
A procedure, based on the complex formation of malachite green with phosphomolybdate under acidic conditions, to measure inorganic orthophosphate in the nanomolar range is described. The addition of polyvinyl alcohol is required to stabilize the dye-phosphomolybdate complex. The advantages of
K W Harder et al.
The Biochemical journal, 298 ( Pt 2), 395-401 (1994-03-01)
The intracellular domain of human protein tyrosine phosphatase beta (HPTP beta) (44 kDa) was expressed in bacteria, purified using epitope 'tagging' immunoaffinity chromatography, and characterized with respect to kinetic profile, substrate specificity and potential modulators of enzyme activity. A chromogenic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service