Skip to Content
Merck
All Photos(1)

Key Documents

17-313

Sigma-Aldrich

PP2A Immunoprecipitation Phosphatase Assay Kit

Synonym(s):

PP2A Activity Assay Kit, Phosphatase Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

25 ASSAYS
€658.00

€658.00


Please contact Customer Service for Availability


Select a Size

Change View
25 ASSAYS
€658.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

€658.00


Please contact Customer Service for Availability

description

Phosphatase Assay

Quality Level

product line

Upstate®

Application

PP2A Immunoprecipitation Phosphatase Assay Kit can be used to detect purified PP2A or PP2A from a cellular lysate. It utilizes the Malachite Green & contains components that have been matched to optimize sensitivity & range of detection.

Quality

Routinely evaluated by immunoprecipitation of purified PP2A (14-111) or PP2A from a cellular lysate. The activity is assessed by dephosphorylation of the phosphopeptide (K-R-pT-I-R-R).

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service