Skip to Content
Merck
All Photos(1)

Key Documents

1.50618

Supelco

Pressure Relief Valve

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
Pricing and availability is not currently available.

technique(s)

ion chromatography: suitable

Quality Level

General description

SeQuant® CARS (Continuous Anion Regeneration System) with SAMS (SeQuant Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.

This pressure relief valve protects your SAMS suppressor from breaking at high backpressures and is recommended for new installations.
back-pressure guard valve 100 psi for SAMS suppressors SeQuant

Analysis Note

visual control
Appearance: conforms

Legal Information

SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service