Skip to Content
Merck
All Photos(1)

Key Documents

1.46815

Millipore

Mossel Broth

Selective enrichment of Enterobacteriaceae from non-sterile pharmaceutical products, foodstuff, and other materials, pkg of (1 L in 1 L Bottle with blue screw cap and 3 loci (6 bottles per box))

Synonym(s):

Buffered Glucose-Brilliant Green Bile Broth, EE Broth, Enterobacteriaceae Enrichment Broth

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 1 L
€115.00

€115.00


Estimated to ship on31 March 2025Details


Request more information

Select a Size

Change View
6 X 1 L
€115.00

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

€115.00


Estimated to ship on31 March 2025Details


Request more information

Agency

EP
JP
USP

Sterilization Compatibility

autoclavable compatible

form

liquid

packaging

pkg of (1 L in 1 L Bottle with blue screw cap and 3 loci (6 bottles per box))

technique(s)

microbiological culture: suitable

pH

7.0-7.4 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

selective for enterobacteriaceae

General description

Mossel Broth is a selective enrichment media for bile-tolerant, Gram-negative bacteria, and particularly Enterobacteriaceae such as Salmonella, Shigella, and other enteric pathogens from non-sterile pharmaceutical products, foodstuff, and other materials. Glucose is the main energy source and supports the rapid growth of all Enterobacteriaceae. The enzymatic digest of animal tissue provides nitrogen, vitamins, and amino acids. The high buffering capacity of the culture medium prevents the acid formed from killing the culture. The undesired accompanying bacterial flora is completely inhibited by brilliant green and ox bile.

Application

Mossel Broth is designed for selective enrichment of bile-tolerant, Gram-negative bacteria, particularly Enterobacteriaceae such as Salmonella, Shigella and other enteric pathogens from non-sterile pharmaceutical products, foodstuff, and other materials.

Analysis Note

The medium after preparation appears clear and medium green.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service