Skip to Content
Merck
All Photos(1)

Key Documents

1.46023

Millipore

HEIMPLATE Mannitol Salt Agar

Heimplate, plate diam. 90 mm, ICR Settle plate for long incubation in non-sterile pharmaceuticals -, for Staphylococcus spp.

Synonym(s):

MSA, Chapman Medium

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
€52.60

€52.60


Estimated to ship on28 March 2025Details


Request more information

Select a Size

Change View
20 PKG
€52.60

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

€52.60


Estimated to ship on28 March 2025Details


Request more information

Agency

EP
JP
USP

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use
ICR Settle plate for long incubation in non-sterile pharmaceuticals -

packaging

pack of 20-120 plates

manufacturer/tradename

Heimplate

technique(s)

microbiological culture: suitable

plate diam.

90 mm

plate filling volume

30 mL

pH

7.4 ( in H2O)

application(s)

microbiology
pharmaceutical
surface monitoring

storage temp.

15-25°C

suitability

Staphylococcus spp.

General description

Mannitol Salt Agar is a modified version of the selective agar proposed by Chapman (1945) for the isolation and presumptive identification of Staphylococcus aureus. The medium supports the growth of Staphylococcus aureus, whereas its high salt content (7.5%) inhibits the growth of many other microorganisms. In contrast to most other Staphylococci, coagulase-positive S. aureus can build acids from mannitol. Therefore, they build up yellow colonies with yellow zones. Beef extract, a pancreatic digest of casein, and animal tissue in the media provide carbon, nitrogen compounds, long-chain amino acids, vitamins, trace nutrients and other essential growth factors. Phenol red indicator aids in the detection of acid produced by fermentation of mannitol.

Application

The Mannitol Salt Agar media is recommended for the isolation and presumptive identification of Staphylococcus aureus in non-sterile pharmaceuticals and foods. However, the coagulase test will need to confirm the findings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service