Skip to Content
Merck
All Photos(1)

Key Documents

1.46767

Millipore

ReadyPlate 55 Cetrimide Nalidixic Acid Agar

To detect Pseudomonas aeruginosa, diam. 55 mm, box of 20 plates

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
€41.00

€41.00


Estimated to ship on24 March 2025Details


Request more information

Select a Size

Change View
20 EA
€41.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

€41.00


Estimated to ship on24 March 2025Details


Request more information

Agency

ISO 16266

Quality Level

form

plate (ready-to-use)

feature

ready-to-use

packaging

box of 20 plates

manufacturer/tradename

ReadyPlate

diam.

55 mm

application(s)

filtration
food and beverages
microbiology

storage temp.

15-25°C

suitability

Pseudomonas aeruginosa

General description

ReadyPlate 55 agar plates are stackable specially designed plates that helps increase the safety during handling, transport, incubation and comes in a lockable format. ReadyPlate 55 agar plates come with a diameter of 55 mm. Cetrimide Nalidixic acid Agar is used for the isolation and enumeration of Pseudomonas aeruginosa from food and water samples. Gelatine peptone and Casein hydrolysate provides amino acids and other complex nitrogenous substances. In the presence of Cetrimide (a selective agent which acts as a quaternary ammonium cationic detergent), bacterial cells growth other than P. aeruginosa is inhibited due to the release of nitrogen and phosphorous. Nalidixic acid present in the supplement also improves the inhibition of the accompanying microbial flora. The amount of potassium sulphate and magnesium chloride enhances the pigment production of Pseudomonas aeruginosa.

Application

ReadyPlate 55 Cetrimide Nalidixic acid Agar is used for the isolation and enumeration of Pseudomonas aeruginosa by membrane filtration method of water.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service