Skip to Content
Merck
All Photos(2)

Key Documents

1.46002

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

Sodium thiosulfate, Tween®, Lecithin, Histidine, Plate for Long incubation, aseptically filled, plate diam. 90 mm, suitable for air monitoring

Synonym(s):

CASO Agar, Casein Soybean Digest Agar, TSA, Tryptic Soy Agar Settle Plate

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
€52.90
120 PKG
€321.00

€52.90


Estimated to ship on03 April 2025Details


Request more information

Select a Size

Change View
20 PKG
€52.90
120 PKG
€321.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

€52.90


Estimated to ship on03 April 2025Details


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

aseptically filled

form

plate (ready-to-use)

feature

ready-to-use
Plate for Long incubation

composition

Histidine
Lecithin
Sodium thiosulfate
Tween®

plate diam.

90 mm

pH

7.3 ( in H2O)

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

15-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar + LTHTh in 90 mm settle plates with 30 mL filling volume for long incubation is designed for the detection of the total aerobic microbial count in the air via active or passive air monitoring as well as fingerprints of personnel in controlled environments.
TSA medium is used for the cultivation and isolation of a wide range of bacteria, yeasts, and molds. The formulation contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium.The medium is supplemented with neutralizers such as lecithin, polysorbate (Tween®) 80, histidine, and sodium thiosulphate to inactivate disinfectant residues.

Application

Tryptic Soy Agar + LTHTh - in 90 mm settle plates is designed for the determination of the total aerobic microbial count in the air via active or passive air monitoring as well as fingerprints of personnel in controlled environments.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Related Content

Settle plates for isolators and critical cleanrooms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service