Skip to Content
Merck
All Photos(1)

Key Documents

1.46758

Millipore

Chromocult® Coliform Agar Kit

ReadyPlate, 55mm plates with EZPAK 0.45 white gridded membrane for the simultaneous enumeration of coliform and E. coli using the Membrane Filtration method

Synonym(s):

CCA, Chromogenic Coliform Agar

Sign Into View Organizational & Contract Pricing

Select a Size

150 EA
€383.00

€383.00


In StockDetails


Request more information

Select a Size

Change View
150 EA
€383.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

€383.00


In StockDetails


Request more information

material

gridded white mixed cellulose esters (MCE) membrane

Quality Level

Agency

ISO 9308

form

plate (ready-to-use)

shelf life

9 mo.

feature

ready-to-use

manufacturer/tradename

ReadyPlate

packaging

pkg of (150 x 55mm Plates and 150 membranes)

application(s)

filtration
food and beverages
microbiology

storage temp.

15-25°C

suitability

Escherichia coli
coliforms

General description

Chromocult® Coliform Agar Kit contains:
ReadyPlate CCA plates of 55mm diameter is a selective and differential chromogenic culture medium. It is a standard medium in water testing for simultaneous detection and enumeration of coliform and E. coli by the combination of two chromogenic substrates. The interaction of selected peptones, pyruvate, sorbitol, and phosphate buffer promotes rapid colony growth, even for sub-lethally injured coliform bacteria. The growth of Gram-positive bacteria, as well as certain Gram-negative bacteria, is inhibited by the presence of Tergitol 7, with no negative effect on the growth of coliform bacteria. E. coli cleaves both Salmon-GAL and X-glucuronide so that positive colonies take on a dark-blue to violet color. These are easily distinguished from other coliform colonies, which have a pink to red color.
EZ-Pak Filters are ready-to-use, sterile filters made of Mixed cellulose esters (MCE) with 0.45 μm pore size, white, gridded, and 50mm diameter. These filters possess high recoveries.

Application

Chromocult® Coliform Agar Kit contains 55mm plates with EZPAK 0.45 μm white gridded membrane for the simultaneous enumeration of coliform and E.coli using the Membrane Filtration method in drinking water, waters with low bacterial background flora.

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany
TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Related Content

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service