Skip to Content
Merck
All Photos(2)

Key Documents

1.00850

Millipore

Chromocult® Coliform Agar ES

suitable for microbiology

Synonym(s):

Coliform Agar, Coliform agar ES, CCA ES

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€818.00

€818.00


Estimated to ship on30 June 2025Details


Request more information

Select a Size

Change View
500 G
€818.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106213
NACRES:
NA.74

€818.00


Estimated to ship on30 June 2025Details


Request more information

sterility

non-sterile

Quality Level

form

granular

manufacturer/tradename

ChromoCult®

technique(s)

microbiological culture: suitable

pH

6.8-7.2 (25 °C, 34.6 g/L in H2O)

solubility

34.6 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Escherichia coli
coliforms

Looking for similar products? Visit Product Comparison Guide

General description

Chromocult® Coliform Agar ES is a selective and differential chromogenic culture medium intended for use in microbiology laboratories analyzing food and animal feeds. The modified Chromocult® Coliform agar with enhanced selectivity allows the simultaneous detection of coliforms and E. coli in samples with high bacterial bioburden. A combination of suitable peptones and buffering with MOPS encourages rapid growth of coliforms and optimum transformation of the chromogenic substrates. The bile salts and propionate inhibit the growth of Gram-positive and Gram-negative accompanying flora. Simultaneous detection of total coliforms and E. coli is achieved by using a combination of two chromogenic substrates. The substrate Salmon-β-D-GAL is cleaved by β-D-galactosidase, characteristic for coliforms, resulting in a salmon to red coloration of coliform colonies. The substrate X-β-D-glucuronide is cleaved via β-D-glucuronidase, characteristic for E. coli, causing positive colonies to turn blue. As E. coli cleaves Salmon-β-D-GAL as well as X-β-D-glucuronide, its colonies turn dark violet and are easily differentiated from the other coliforms, these being salmon-red.

Application

Chromocult® Coliform Agar is a selective agar suitable for the simultaneous detection and colony count of total coliforms and E. coli in foods. This medium enables E. coli and coliforms from food matrices such as raw ground beef, raw ground chicken, and raw milk to be detected, differentiated, and enumerated within 24 hours.

Analysis Note

Appearance (clearness): clear
pH-value (25 °C): 6.8 - 7.2
Recovery on test medium (Escherichia coli ATCC 11775 (WDCM 00090)): ≥ 70 %
Recovery on test medium (Citrobacter freundii ATCC 8090): ≥ 70 %
Recovery on test medium (Enterobacter aerogenes ATCC 13048): ≥ 70 %
Recovery on test medium (Proteus mirabilis ATCC 29906 (WDCM 00023)):
Growth (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): inhibited
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): inhibited
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): inhibited
Growth (Bacillus subtilis ATCC 6633 (WDCM 00003)): inhibited
Colony colour (Escherichia coli ATCC 11775 (WDCM 00090)): blue
Colony colour (Citrobacter freundii ATCC 8090): pink-red
Colony colour (Enterobacter aerogenes ATCC 13048): pink-red
Incubation: 24 hrs.; 35 °C; aerobic.
Reference: Tryptic soy agar.
Inhibited strains tested with three-loop-streak.

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service